2,3-Dimethylbenzoylchlorid

Lieferant: Thermo Scientific

TL00220.0005 TL00220.10 TL00220.50
MAYBTL00220.0005EA 0 EUR
MAYBTL00220.0005 MAYBTL00220.10 MAYBTL00220.50
2,3-Dimethylbenzoylchlorid
2,3-Dimethylbenzoylchlorid
Formel: C₉H₉ClO
MDL: MFCD00045217
CAS-Nummer: 21900-46-9
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR