3-Ethyl-2-iodtoluol

Lieferant: Thermo Scientific

TL00249.1
MAYBTL00249.1EA 0 EUR
MAYBTL00249.1
3-Ethyl-2-iodtoluol
3-Ethyl-2-iodtoluol
Formel: C₉H₁₁I
MDL: MFCD00052855
CAS-Nummer: 175277-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR