4'-Nitro-2'-(trifluormethyl)acetanilid

Lieferant: Thermo Scientific

TL00253.1
MAYBTL00253.1EA 0 EUR
MAYBTL00253.1
4'-Nitro-2'-(trifluormethyl)acetanilid
4'-Nitro-2'-(trifluormethyl)acetanilid
Formel: C₉H₇F₃N₂O₃
MDL: MFCD00052846
CAS-Nummer: 395-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR