4'-Cyan-2'-(trifluormethyl)acetanilid

Lieferant: Thermo Scientific

TL00263DA TL00263.10
MAYBTL00263DAEA 0 EUR
MAYBTL00263DA MAYBTL00263.10
4'-Cyan-2'-(trifluormethyl)acetanilid
4'-Cyan-2'-(trifluormethyl)acetanilid
Formel: C₁₀H₇F₃N₂O
MDL: MFCD00052844
CAS-Nummer: 175277-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR