2-(Trifluormethyl)benzamid

Lieferant: Thermo Scientific

Synonyme: 2-(Trifluormethyl)benzoesäureamid

TL00337.50
MAYBTL00337.50EA 0 EUR
MAYBTL00337.50
2-(Trifluormethyl)benzamid
2-(Trifluormethyl)benzamid
Formel: C₈H₆F₃NO
Molecular Weight: 189.14 g/mol
MDL: MFCD00014798
CAS-Nummer: 360-64-5
EINECS: 206-637-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR