1-Iod-2-(trifluormethoxy)benzol

Lieferant: Thermo Scientific

TL00358.1 TL00358.10 TL00358EE
MAYBTL00358.1EA 0 EUR
MAYBTL00358.1 MAYBTL00358.10 MAYBTL00358EE
1-Iod-2-(trifluormethoxy)benzol
1-Iod-2-(trifluormethoxy)benzol
Formel: C₇H₄F₃IO
MDL: MFCD00042410
CAS-Nummer: 175278-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR