2-(Trifluormethoxy)benzonitril

Lieferant: Thermo Scientific

TL00362.50
MAYBTL00362.50EA 0 EUR
MAYBTL00362.50
2-(Trifluormethoxy)benzonitril
2-(Trifluormethoxy)benzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187,12 g/mol
Siedepunkt: 80…82 °C (15 mmHg)
MDL: MFCD00042409
CAS-Nummer: 63968-85-4
EINECS: 264-584-0
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR