1-Nitro-4-(trifluormethoxy)benzol

Lieferant: Thermo Scientific

TL00370.1 TL00370.10
MAYBTL00370.1EA 0 EUR
MAYBTL00370.1 MAYBTL00370.10
1-Nitro-4-(trifluormethoxy)benzol
1-Nitro-4-(trifluormethoxy)benzol
Formel: C₇H₄F₃NO₃
MDL: MFCD00040302
CAS-Nummer: 713-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR