α-Chlor-4-(trifluormethoxy)toluol

Lieferant: Thermo Scientific

TL00386DA TL00386EA
MAYBTL00386DAEA 0 EUR
MAYBTL00386DA MAYBTL00386EA
α-Chlor-4-(trifluormethoxy)toluol
α-Chlor-4-(trifluormethoxy)toluol
Formel: C₈H₆ClF₃O
MDL: MFCD00052326
CAS-Nummer: 65796-00-1
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR