α-Chlor-4-(trifluormethoxy)toluol
Lieferant: Thermo Scientific
Synonyme:
4-(Trifluormethoxy)benzylchlorid, p-(Trifluormethoxy)benzylchlorid
TL00386DA
TL00386EA
MAYBTL00386DAEA
0
EUR
MAYBTL00386DA
MAYBTL00386EA
α-Chlor-4-(trifluormethoxy)toluol
α-Chlor-4-(trifluormethoxy)toluol
Formel:
C₈H₆ClF₃O Molecular Weight: 210.58 |
MDL:
MFCD00052326 CAS-Nummer: 65796-00-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...