4-(Trifluormethoxy)phenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 4-(Trifluoromethoxy)phenylisothiocyanate

TL00411DA TL00411EA
MAYBTL00411DAEA 0 EUR
MAYBTL00411DA MAYBTL00411EA
4-(Trifluormethoxy)phenylisothiocyanat
4-(Trifluormethoxy)phenylisothiocyanat
Formel: C₈H₄F₃NOS
Molecular Weight: 219.19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00042412
CAS-Nummer: 64285-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR