4-(Trifluormethoxy)phenylisothiocyanat
Lieferant: Thermo Scientific
Synonyme:
4-(Trifluoromethoxy)phenylisothiocyanate
TL00411DA
TL00411EA
MAYBTL00411DAEA
0
EUR
MAYBTL00411DA
MAYBTL00411EA
4-(Trifluormethoxy)phenylisothiocyanat
4-(Trifluormethoxy)phenylisothiocyanat
Formel:
C₈H₄F₃NOS Molecular Weight: 219.19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00042412 CAS-Nummer: 64285-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...