Methyl-4-(trifluormethoxy)benzoat

Lieferant: Thermo Scientific

TL00423DA TL00423.10
MAYBTL00423DAEA 0 EUR
MAYBTL00423DA MAYBTL00423.10
Methyl-4-(trifluormethoxy)benzoat
Methyl-4-(trifluormethoxy)benzoat
Formel: C₉H₇F₃O₃
Molecular Weight: 220,15 g/mol
MDL: MFCD00041006
CAS-Nummer: 780-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR