4-(Trifluormethyl)phenylisothiocyanat
Lieferant: Thermo Scientific
Synonyme:
α,α,α-Trifluor-p-tolylisothiocyanat
TL00445.1
TL00445EA
MAYBTL00445.1EA
0
EUR
MAYBTL00445.1
MAYBTL00445EA
4-(Trifluormethyl)phenylisothiocyanat
4-(Trifluormethyl)phenylisothiocyanat
Formel:
C₈H₄F₃NS Siedepunkt: 205…207 °C (1013 hPa) Schmelzpunkt: 29…31 °C |
MDL:
MFCD00039645 CAS-Nummer: 1645-65-4 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...