4-(Difluormethoxy)benzaldehyd

Lieferant: Thermo Scientific

TL00446DA TL00446.10
MAYBTL00446DAEA 0 EUR
MAYBTL00446DA MAYBTL00446.10
4-(Difluormethoxy)benzaldehyd
4-(Difluormethoxy)benzaldehyd
Formel: C₈H₆F₂O₂
MDL: MFCD00042252
CAS-Nummer: 73960-07-3
EINECS: 277-653-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR