2-Brom-4'-(difluormethoxy)acetophenon

Lieferant: Thermo Scientific

Synonyme: 4-(Difluoromethoxy)phenacyl bromide

TL00461DA TL00461EA
MAYBTL00461DAEA 0 EUR
MAYBTL00461DA MAYBTL00461EA
2-Brom-4'-(difluormethoxy)acetophenon
2-Brom-4'-(difluormethoxy)acetophenon
Formel: C₉H₇BrF₂O₂
MDL: MFCD00052200
CAS-Nummer: 141134-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR