1-(Difluormethoxy)-4-nitrobenzol

Lieferant: Thermo Scientific

TL00492.1 TL00492.10
MAYBTL00492.1EA 0 EUR
MAYBTL00492.1 MAYBTL00492.10
1-(Difluormethoxy)-4-nitrobenzol
1-(Difluormethoxy)-4-nitrobenzol
Formel: C₇H₅F₂NO₃
MDL: MFCD00085009
CAS-Nummer: 1544-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR