4-(Difluormethoxy)iodbenzol

Lieferant: Thermo Scientific

TL00517.1 TL00517.10
MAYBTL00517.1EA 0 EUR
MAYBTL00517.1 MAYBTL00517.10
4-(Difluormethoxy)iodbenzol
4-(Difluormethoxy)iodbenzol
Formel: C₇H₅F₂IO
MDL: MFCD00085008
CAS-Nummer: 128140-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR