4-(Difluormethoxy)iodbenzol
Lieferant: Thermo Scientific
TL00517.1
TL00517.10
MAYBTL00517.1EA
0
EUR
MAYBTL00517.1
MAYBTL00517.10
4-(Difluormethoxy)iodbenzol
4-(Difluormethoxy)iodbenzol
Formel:
C₇H₅F₂IO |
MDL:
MFCD00085008 CAS-Nummer: 128140-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...