4-Iod-3-nitrotoluol

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Iodo-4-methyl-2-nitrobenzene

TL00543.50
MAYBTL00543.50EA 0 EUR
MAYBTL00543.50
4-Iod-3-nitrotoluol
4-Iod-3-nitrotoluol
Formel: C₇H₆INO₂
Molecular Weight: 263,03 g/mol
Schmelzpunkt: 55 °C
MDL: MFCD00024196
CAS-Nummer: 5326-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR