4'-(Trifluormethoxy)acetanilid

Lieferant: Thermo Scientific

TL00595.1 TL00595.50
MAYBTL00595.1EA 0 EUR
MAYBTL00595.1 MAYBTL00595.50
4'-(Trifluormethoxy)acetanilid
4'-(Trifluormethoxy)acetanilid
Formel: C₉H₈F₃NO₂
MDL: MFCD00075293
CAS-Nummer: 1737-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR