4-Brom-2-(trifluormethoxy)anilin
Lieferant: Thermo Scientific
TL00627.1
TL00627EA
TL00627.50
MAYBTL00627.1EA
0
EUR
MAYBTL00627.1
MAYBTL00627EA
MAYBTL00627.50
4-Brom-2-(trifluormethoxy)anilin
4-Brom-2-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO |
MDL:
MFCD00179338 CAS-Nummer: 175278-09-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...