2-Chlor-4-iodtoluol

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-4-iodo-1-methylbenzene, 1-Chloro-5-iodo-2-methylbenzene

TL00633.1 TL00633.10 TL00633.50
MAYBTL00633.1EA 0 EUR
MAYBTL00633.1 MAYBTL00633.10 MAYBTL00633.50
2-Chlor-4-iodtoluol
2-Chlor-4-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252.48 g/mol
MDL: MFCD00040872
CAS-Nummer: 83846-48-4
EINECS: 281-024-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR