3,4-Difluorbenzonitril
Lieferant: Thermo Scientific
TL00656DA
TL00656EA
TL00656.50
MAYBTL00656DAEA
0
EUR
MAYBTL00656DA
MAYBTL00656EA
MAYBTL00656.50
3,4-Difluorbenzonitril
3,4-Difluorbenzonitril
Formel:
C₇H₃F₂N Siedepunkt: 189 °C (1013 hPa) Schmelzpunkt: 51 °C |
MDL:
MFCD00011666 CAS-Nummer: 64248-62-0 EINECS: 264-751-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...