Methyl-3-methyl-4-nitrobenzoat
Lieferant: Thermo Scientific
Synonyme:
Methyl 3-methyl-4-nitrobenzoate
TL00715DA
TL00715EE
MAYBTL00715DAEA
0
EUR
MAYBTL00715DA
MAYBTL00715EE
Methyl-3-methyl-4-nitrobenzoat
Methyl-3-methyl-4-nitrobenzoat
Formel:
C₉H₉NO₄ Molecular Weight: 195,18 g/mol |
MDL:
MFCD00085640 CAS-Nummer: 24078-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...