4-Chlor-2-methylphenylisothiocyanat

Lieferant: Thermo Scientific

TL00725.50
MAYBTL00725.50EA 0 EUR
MAYBTL00725.50
4-Chlor-2-methylphenylisothiocyanat
4-Chlor-2-methylphenylisothiocyanat
Formel: C₈H₆ClNS
Molecular Weight: 183,66 g/mol
Siedepunkt: 136 °C (13 mmHg)
Schmelzpunkt: 36 °C
MDL: MFCD00041089
CAS-Nummer: 23165-53-9
EINECS: 245-472-0
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR