2-Fluor-4-nitrobenzoesäure

Lieferant: Thermo Scientific
Achtung

TL00808DA TL00808EA
MAYBTL00808DAEA 0 EUR
MAYBTL00808DA MAYBTL00808EA
2-Fluor-4-nitrobenzoesäure
2-Fluor-4-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185.11 g/mol
MDL: MFCD00275565
CAS-Nummer: 403-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR