6-Amino-3-brom-2-methylpyridin

Lieferant: Thermo Scientific
Achtung

XBX00110.1 XBX00110EA
MAYBXBX00110.1EA 0 EUR
MAYBXBX00110.1 MAYBXBX00110EA
6-Amino-3-brom-2-methylpyridin
6-Amino-3-brom-2-methylpyridin
Formel: C₆H₇BrN₂
MDL: MFCD00068230
CAS-Nummer: 42753-71-9
EINECS: 255-927-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR