Tissue Marking Dye Kit, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

MDT100-1KT
MDT100-1KTEA 917 EUR
MDT100-1KT
Tissue Marking Dye Kit, Sigma-Aldrich®
Tissue Marking Dye Kit

Designed for orientation of excised surgical specimens. This method of permanently marking the margins of tissue specimens is very beneficial to surgeon and pathologists testing for skin cancer.


  • Suitable for haematology
  • Suitable for histology
  • 5 colors


This method of marking multiple margins of specimens was originally documented by Dr. Frederick Mohs. Five 2 oz. bottles of dye: blue, black, yellow, red and green. Convenient holder and application sticks.


Kit component only
Red Dye (R3780) 2 oz
Green Dye (G4421) 2 oz
Blue Dye (B8186) 2 oz
Yellow Dye (Y0146) 2 oz
Black Dye (B8311) 2 oz

Lagertemperatur: Kühlschrank
UN: 3316
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR