DS2 Serum-free/protein-free medium, 1X without L-glutamine, Corning® insectagro®
Lieferant: Corning
13-402-CV
MDTC13-402-CVEA 248.82 EUR
MDTC13-402-CV
DS2 Serum-free/protein-free medium, 1X without L-glutamine, Corning® insectagro®
Zellkulturmedien
Corning® insectagro® DS2 Serum-Free/Protein-Free Medium was developed for the growth and maintenance of Drosophila Schneider 2 (S2) cells to be used in heterologous protein expression
At the optimal temperature range 22-24ºC, DS2 cells grow as a loose monolayer and are readily adaptable to growth in suspension. Under these conditions, the cells require minimal adaptation to serum-free culture.