Trace elements B, Corning®
Lieferant: Corning
25-022-CI
MDTC25-022-CIEA 59.78 EUR
MDTC25-022-CI
Trace elements B, Corning®
Medien für die Mikrobiologie Additive und Supplemente für die Mikrobiologie
Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
The combination of Trace Elements B contains the trace elements of MCDB 104 and of MCDB 301.
This product is proprietary and not available from other major suppliers.