HT (hypoxanthine, thymidine) media, cellgro™, Corning®
Lieferant: Corning
25-047-CI
MDTC25-047-CIEA
31.53
EUR
MDTC25-047-CI
HT (hypoxanthine, thymidine) media, cellgro™, Corning®
Zellkulturmedien
HT media is a 'rescue media' that provides pre-formed purines and pyrimidines to overcome the effects of residual intracellular aminopterin.
- Complete Certificate of Analysis available for each production lot, along with full formulation information
- Produced under the highest industry standards to ensure superior results