Wasser, Cell Culture grade, tested USP Injection, sterile for sterile Water for Injection , Corning®
Lieferant: Corning
Synonyme:
Wasserstoffoxid, Hydrogenoxid
Corning® Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications. Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications.
- Complete Certificate of Analysis available for each production lot
- Produced under the highest industry standards to ensure superior results
- Meets WFI Standards
Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 Schmelzpunkt: 0 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Mycoplasma | Tested negative |
Plating efficiency | Pass |
Oxidizable substances Solution | remains faintly pink |
Total organic carbon (before packaging) USP <643> | < 500 ppb |
Water conductivity (packaged water) USP <645> | Meets USP Requirements |
Water conductivity (before packaging) USP <645> | Meets USP temperature/conductivity requirements |
Endotoxin USP <85> | ≤ 0.025 EU/mL |
Sterility USP <71> | Pass |
Particulate matter USP <788> |
≤ 25 particles/mL that are ≥ 10 μm in size ≤ 3 particles/mL that are ≥ 25 μm in size |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...