Amphotericin B 250 µg/ml

Lieferant: Corning

Synonyme: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

30-003-CF
MDTC30-003-CFEA 157.74 EUR
MDTC30-003-CF
Amphotericin B 250 µg/ml
Amphotericin B

This antimycotic interferes with fungal membrane permeability by forming channels in cell membranes, causing small molecules to leak from the cell. It is recommended for use in cell culture applications at 10 ml/L and is stable at 37 ºC for 3 days.


Verpackung: Shipped frozen.

Formel: C₄₇H₇₃NO₁₇
Molecular Weight: 924,08 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00877763
CAS-Nummer: 1397-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR