Fetal Bovine Serum, Regular, USDA Approved Origin
Lieferant: Corning
35-011-CV
MDTC35-011-CVEA 660 EUR
MDTC35-011-CV
Fetal Bovine Serum, Regular, USDA Approved Origin
Seren
Wide variety of fetal bovine serum (FBS) options to satisfy most common applications and is a perfect complement to Corning classical cell culture media. Products are evaluated for quality, performance
and analytical testing, including sterility, mycoplasma, virus testing, total protein, endotoxin and hemoglobin levels.
  • Storage temperature is −40 to −10 °C
Standards: ISIA, CEP, EDOM certified.