Bovine calf serum, 1 L, iron-fortified, United States origin, Corning®

Lieferant: Corning
35-053-CM
MDTC35-053-CMEA 227.79 EUR
MDTC35-053-CM
Bovine calf serum, 1 L, iron-fortified, United States origin, Corning®
Seren
Bovine calf serum, United States origin are collected at government approved abattoirs in the United States.

  • Cattle are less than 35 weeks of age at time of collection
  • True pooled and triple 0,1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Shelf life: 60 months
  • Format: Liquid

Iron fortified to raise iron saturation to between 70 - 80% of total iron binding capacity.

Standards: Granted a Certificate of Suitability, R0-CEP 2018-305, by the European Directorate for the Quality of Medicines (EDQM).
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR