Corning® Gamma-Irradiated FBS, Corning
Lieferant: Corning
35-015-CV 35-016-CV 35-070-CV
MDTC35-015-CVEA 695.99 EUR
MDTC35-015-CV MDTC35-016-CV MDTC35-070-CV
Corning® Gamma-Irradiated FBS, Corning
Seren
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds
  • Heat-inactivated Fetal Bovine Serum (FBS) is heated at 56°C for 30 minutes