Wasser, Reagenzqualität, entspricht den analytischen Spezifikationen von USP, steril

Lieferant: Corning

Synonyme: Wasserstoffoxid, Hydrogenoxid

46-002-LF
MDTC46-002-LFEA 70.9 EUR
MDTC46-002-LF
Wasser, Reagenzqualität, entspricht den analytischen Spezifikationen von USP, steril
Wasser

Reagent grade water is ideal for the preparation of reagents, rinsing glassware and plastic ware, cleaning electrodes and other common laboratory tasks requiring highly pure water.


  • No toxic agents, including DEPC, are used in its preparation
  • RNAse, DNAse and Protease free


The design allows for portability, ease of storage and convenient dispensing. Upon packaging, the water meets USP conductivity requirements for water for injection (WFI) and is RNAse-, DNAse- and Protease-free. Provided in packs of 2 x 4L cubes.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR