10X Tris buffered saline, liquid, Corning®
Lieferant: Corning
46-012-CM
MDTC46-012-CMEA
338.08
EUR
MDTC46-012-CM
10X Tris buffered saline, liquid, Corning®
Puffer
Gewebekultur-Puffer
Tris Buffered Saline (TBS) is an isotonic, non-toxic buffer used in some biomedical techniques to maintain pH within a relatively narrow range.
TBS can shift pH relatively dramatically as temperature changes, with the pKa declining approximately 0,03 units per degree Celsius rise in temperature.