X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid), Pulver, Corning®

Lieferant: Corning

46-101-RF
MDTC46-101-RFEA 337.45 EUR
MDTC46-101-RF
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid), Pulver, Corning®
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)

X-Gal (5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside) is a substrate of b-galactosidase, a gene product of the lac operon in E. coli.


X-Gal included in bacterial medium will be hydrolyzed by ß-galactosidase to produce indoxyl, a colormetric indicator of the activity of the lac operon.

Formel: C₁₄H₁₅BrClNO₆
Schmelzpunkt: 237…239 °C
MDL: MFCD00005666
CAS-Nummer: 7240-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR