Yeast extract, powder, Corning®
Lieferant: Corning
61-231-RO
MDTC61-231-ROEA 265.81 EUR
MDTC61-231-RO
Yeast extract, powder, Corning®
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates obtained from autolysed yeast
This is the water soluble portion with intact B-complex vitamins and can be used as an additive for culture media.