Neomycinsulfat, Pulver

Lieferant: Corning

Synonyme: Neomycin Sulfatsalz

61-241-RG
MDTC61-241-RGEA 81.03 EUR
MDTC61-241-RG
Neomycinsulfat, Pulver
Neomycinsulfat

Neomycin, a nucleoside antibiotic and aminoglycoside, is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3'-terminal end of aminoacyl-tRNA


It prevents growth of bacteria, protozoa, algae, and mammalian cells. Organisms carrying the resistance gene puromycin acetyltransferase are effectively protected from this compound. Neomycin is effective against Gram-negative and Gram-positive bacteria.


Verpackung: Shipping temperature: Ambient

Formel: C₂₃H₄₆N₆O₁₃·3H₂SO₄
Molecular Weight: 908,87 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150827
CAS-Nummer: 1405-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR