Corning® Human recombinant albumin (from serum)
Lieferant: Corning
62-450-RF
MDTC62-450-RFEA
80.4
EUR
MDTC62-450-RF
Corning® Human recombinant albumin (from serum)
Proteine und Peptide
Corning® cellgro® Albumin is a chemically-defined recombinant human serum albumin (rhHSA). Having an essential and non-essential fatty acid profile, this is a powerful media supplement that outperforms plasma-derived human serum albumin (pHSA), bovine serum albumin (BSA) and FBS. Some prominent applications for cellgro® rhAlbumin are as a supplement for CHO cell culture and for the culture of various other cell lines. This product is plant-derived and completely animal-origin free.