Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Saturated Straight Chains
Lieferant: Merck
ME10-1KT
ME10-1KTEA 790 EUR
ME10-1KT
Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Saturated Straight Chains
Standards Organische Standards
  • GHS signal: GHS02, GHS05
Lieferumfang: Kit Components: Caproic (C6:0) 1 g; Caprylic (C8:0) 1 g; Capric (C10:0) 1 g; Lauric (C12:0) 1 g; Myristic (C14:0) 1 g; Palmitic (C16:0) 1 g; Stearic (C18:0) 1 g; Arachidic (C20:0) 1 g; Behenic (C22:0) 1 g; Lignoceric (C24:0) 100 mg.
Bitte beachten P210 - P233 - P240 - P241 - P242 - P273