Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Unsaturated Kit
Lieferant: Merck
ME14-1KT
ME14-1KTEA
837
EUR
ME14-1KT
Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Unsaturated Kit
Standards
Organische Standards
- GHS signal: GHS07
Lieferumfang: Analytes: cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 100 mg; Methyl arachidonate 100 mg; Methyl cis-13-docosenoate 100 mg; Methyl cis-11-eicosenoate 100 mg; Methyl elaidate 100 mg; Methyl linoleate 100 mg; Methyl linolelaidate 100 mg; Methyl linolenate 100 mg; Methyl myristoleate 100 mg; Methyl oleate 100 mg; Methyl palmitoleate 100 mg; Methyl petroselinate 100 mg; Methyl cis-15-tetracosenoate 100 mg.
Bitte beachten P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338