exo-1,3-β-D-Glucanase + β-Glucosidase
Lieferant: MEGAZYME
E-EXBGOS
MEGAE-EXBGOSEA
347
EUR
MEGAE-EXBGOS
exo-1,3-β-D-Glucanase + β-Glucosidase
Enzyme
High purity exo-1,3-β-D-Glucanase (Trichoderma sp.) + β-Glucosidase (Aspergillus niger) for use in research, biochemical enzyme assays and in vitro diagnostic analysis.
- Shipping temperature: Ambient
- Storage temperature: 2 to 8 °C
- Stability: >1 year under recommended storage conditions
- CAZy family: GH3, GH55
- Temperature optima: 40 °C
- Application: For use in the determination of (1,3)(1,6) β-glucan
A wide range of other carbohydrate active enzymes also available.
Specificity:
exo-1,3-β-glucanase: Successive hydrolysis of β-D-glucose units from the non-reducing ends of (1,3)-β-D-glucans, releasing α-glucose.
β-glucosidase: Hydrolysis of terminal, non-reducing β-D-glucosyl residues with release of β-D-glucose.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...