MELAcontrol Pro for Stream Sterilizers
Lieferant: MELAG Medizintechnik OHG
ME01075
MELAME01075EA 467 EUR
MELAME01075
MELAcontrol Pro for Stream Sterilizers
Sterilisationsindikatoren
MELAcontrol Pro with 40 indicator strips is a durable, usable test body for the power-user.
This is a test body for 'Class B' steam sterilizers, which simulate the complicated requirements for sterilizing hollow instruments. This test body is required for batch monitoring of the processing of 'critical B' instruments.
Standards: EN 867-5 standard.
Bestellinformation: The test strips can be re-ordered in a refill package with 250 indicator strips.