Disposable Paper Filter
Lieferant: MELAG Medizintechnik OHG
ME26515
MELAME26515EA 39.5 EUR
MELAME26515
Disposable Paper Filter
Filter Filterpapiere Qualitatives Filterpapier
The sterilisation containers have perforated lids and are equipped with disposable paper filters.
Lieferumfang: It contains 100 disposable paper filters.