4-(Trifluormethylthio)benzylbromid

Lieferant: MENAI

CF1.25 CF1.5
MENACF1.25EA 0 EUR
MENACF1.25 MENACF1.5
4-(Trifluormethylthio)benzylbromid
4-(Trifluormethylthio)benzylbromid
Formel: C₈H₆BrF₃S
MDL: MFCD00129189
CAS-Nummer: 21101-63-3
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR