3-(1,1,2,2-Tetrafluorethoxy)anilin

Lieferant: MENAI

CF7.10
MENACF7.10EA 0 EUR
MENACF7.10
3-(1,1,2,2-Tetrafluorethoxy)anilin
3-(1,1,2,2-Tetrafluorethoxy)anilin
Formel: C₈H₇F₄NO
MDL: MFCD00042457
CAS-Nummer: 831-75-4
EINECS: 212-610-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR